2,4-Toluenediisocyanate ≥80% (remainder 2,6-diisocyanate), tech.

Supplier: Thermo Scientific Chemicals
Danger

36733-30 36733-A1
CAAA36733-30EA 66.99 CAD
CAAA36733-30 CAAA36733-A1
2,4-Toluenediisocyanate ≥80% (remainder 2,6-diisocyanate), tech.
2,4-Toluenediisocyanate

Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate
Toluene diisocyanate is utilized in polyurethane foams and tank trucks. It is also used in concrete sealers for aircraft, sealers and coatings in floor. It is employed as a crosslinking agent for nylon.


Purity: 80% CAS No.: 584-84-9 Molecular Formula: C9H6N2O2 Formula Weight: 174.16 Flash Point: 132° (270°F) Physical Form: Liquid Density: 1.225 Refractive Index: 1.567 Sensitivity: Moisture Sensitive Appearance: Colorless Melting Point: 19.5-21.5° Boiling Point: 251° Beilstein Reference: 744602 Merck Reference: 14,9530 UN No.: 2078 MDL No.: MFCD00002011

Formula: C₉H₆N₂O₂
Boiling Pt: 251 °C
Melting Pt: 19.5…21.5 °C
Density: 1.225
Flash Pt: 132 °C (270 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002011
CAS Number: 584-84-9
EINECS: 209-544-5
UN: 2078
ADR: 6.1,II
Merck Index: 14,9530

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR