Isoamyl alcohol ≥98.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

36716-AP 36716-K2 36716-K7
CAAA36716-APEA 101.12 CAD
CAAA36716-AP CAAA36716-K2 CAAA36716-K7
Isoamyl alcohol ≥98.5% ACS
Isoamyl alcohol

MDL: MFCD00002934 Beilstein Registry No.: 1718835


Purity: 98.5+% CAS No.: 123-51-3 Molecular Formula: (CH3)2CHCH2CH2OH Formula Weight: 88.15 Flash Point: 45°(113°F) Physical Form: Liquid Density: 0.809 Refractive Index: 1.4060 Odor: Unpleasant Melting Point: -117° Boiling Point: 130-132° Beilstein Reference: 1718835 Merck Reference: 14,5195 UN No.: 1105 MDL No.: MFCD00002934

Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130…132 °C
Melting Pt: –117 °C
Density: 0.809
Flash Pt: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3 II
Merck Index: 14,5195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

H2O 0.5 %
Titratable acid 0.002meq/gmeq/g
Evaporation residue 0.003%
Acid and esters (as amyl acetate) 0.2%
Carbonyl (as HCHO) 0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR