Chromium(III) potassium sulfate dodecahydrate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chromium potassium sulphate dodecahydrate, Chromium potassium sulfate dodecahydrate, Chromium (III) potassium bis(sulphate) dodecahydrate, Chromium (III) potassium bis(sulfate) dodecahydrate
MDL: MFCD00149917
Purity: 98.0-102.0%
CAS No.: 7788-99-0
Molecular Formula: CrKO8S2.12H2O
Formula Weight: 499.39 (283.21anhy)
Physical Form: Solid
Density: 1.83
Appearance: Dark purple
Odor: Odorless
Melting Point: 89°
Merck Reference: 14,2229
MDL No.: MFCD00149917
Formula:
CrK(SO₄)₂·12H₂O MW: 499.4 g/mol Melting Pt: 89 Density: 1.83 Storage Temperature: Ambient |
MDL Number:
MFCD00149917 CAS Number: 7788-99-0 EINECS: 233-401-6 Merck Index: 14,2229 |
Specification Test Results
Insoluble matter | 0.01% |
Cl | 0.002% |
Al | 0.02% |
NH3 | 0.01% |
Heavy Metals (as Pb) | 0.01% |
Fe | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...