Starch, soluble ACS for iodometric titrations

Supplier: Thermo Scientific Chemicals

Synonyms: Starch, Vitex® iodine indicator, Amylodextrin

36703-18 36703-30 36703-A1
CAAA36703-18EA 55.14 CAD
CAAA36703-18 CAAA36703-30 CAAA36703-A1
Starch, soluble ACS for iodometric titrations
Starch, soluble

MDL: MFCD00082026 Soluble in hot water


CAS No.: 9005-84-9 Physical Form: Powder Odor: Odorless Melting Point: 256-258° dec. Solubility: Soluble in hot water Merck Reference: 14,8799 MDL No.: MFCD00082026

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Melting Pt: 256…258 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 14,8799

Order Now

Specification Test Results

Solubility Pass test
pH of a 2% solution 5.0-7.0 at 25°
Residue after ignition 0.004
Sensitivity Pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR