trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 97.5-100.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2',2'',2'''-(trans-Cyclohexane-1,2-diylbis(azanetriyl))tetraacetic acid hydrate, DCTA monohydrate, CDTA monohydrate, trans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrate, (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate, 2,2',2'',2'''-[trans-Cyclohexane-1,2-diylbis(azanetriyl)]tetraacetic acid hydrate, CHELATON 4, Titriplex® IV, trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, trans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydrate

36697-06 36697-14 36697-22
CAAA36697-06EA 45.66 CAD
CAAA36697-06 CAAA36697-14 CAAA36697-22
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 97.5-100.5% ACS
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

MDL: MFCD00066429 Beilstein Registry No.: 3222748


Purity: 97.5%-100.5% CAS No.: 125572-95-4 Molecular Formula: C14H22N2O8.H2O Formula Weight: 364.35 (346.34anhy) Physical Form: Solid Melting Point: 213-216° Beilstein Reference: 3222748 MDL No.: MFCD00066429

Formula: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9

Order Now

Specification Test Results

Form Solid
Heavy Metals (as Pb) 0.001%
Fe 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR