Hydrobromic acid 47,0 - 49,0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrogen bromide

36694-AK 36694-K2 36694-M1
CAAA36694-AKEA 108.74 CAD
CAAA36694-AK CAAA36694-K2 CAAAA36694-M1
Hydrobromic acid 47,0 - 49,0% ACS
Hydrogen bromide

MDL: MFCD00011323 Beilstein Registry No.: 3587158 Fieser: 1,450 2,214 3,154 18,180


CAS No.: 10035-10-6 Molecular Formula: HBr Formula Weight: 80.92 Physical Form: Liquid Density: 1.490 Sensitivity: Air Sensitive Appearance: Colorless to yellow Beilstein Reference: 3587158 Merck Reference: 14,4778 UN No.: 1788 MDL No.: MFCD00011323

Formula: HBr
MW: 80.91 g/mol
Boiling Pt: -66.7
Melting Pt: -86
Density: 1.490
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8 II
Merck Index: 14,4778

Order Now

Specification Test Results

Form Liquid
Cl 0.05%
I 0.003%
PO3 0.001%
Sulfate and sulfite (as SO3) 0.003%
Heavy Metals (as Pb) 5 ppm
Fe 1 ppm
Se 0.01 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR