Sodium dichromate dihydrate 99.5-100.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

36692-22 36692-36 36692-A3
CAAA36692-22EA 84.53 CAD
CAAA36692-22 CAAA36692-36 CAAA36692-A3
Sodium dichromate dihydrate 99.5-100.5% ACS
Sodium dichromate dihydrate

MDL: MFCD00149166 Fieser: 1,1059 3,265


Purity: 99.5-100.5% CAS No.: 7789-12-0 Molecular Formula: Cr2Na2O7.2H2O Formula Weight: 298.00 (261.97anhy) Physical Form: Crystalline Density: 2.348 Appearance: Red Odor: Odorless Melting Point: 100° -2H2O Merck Reference: 14,8609 UN No.: 3087 MDL No.: MFCD00149166

Formula: Na₂Cr₂O₇·2H₂O
MW: 298.00 g/mol (261.97 anhy)
Melting Pt: 356.7
Density: 2.348
Storage Temperature: Ambient
MDL Number: MFCD00149166
CAS Number: 7789-12-0
EINECS: 234-190-3
UN: 3087
ADR: 5.1 II
Merck Index: 14,8609

Order Now

Specification Test Results

Insoluble matter 0.005%
Cl 0.005%
SO3 0.01%
Ca 0.003%
MG 0.005%
K 0.01%
Al 0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR