Tin ≥99.5%, granules ACS
Supplier: Thermo Scientific Chemicals
36691-22
36691-36
CAAA36691-22EA
132.72
CAD
CAAA36691-22
CAAA36691-36
Tin ≥99.5%, granules ACS
Tin
MDL: MFCD00133862
Purity: 99.9%
CAS No.: 7440-31-5
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn Boiling Pt: 2270 °C (1013 hPa) Melting Pt: 232 °C Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Specification Test Results
Sb | 0.02% Max. |
As | 1ppm Max. |
Cu | 0.005% Max. |
Fe | 0.01%Max. |
PB | 0.005% Max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...