4-(Dimethylamino)benzaldehyde ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
MDL: MFCD00003381
Beilstein Registry No.: 606802
Notes: Ehrlich's reagent
Slightly soluble in water. Soluble in alcohol ether chloroform acetic acid
Fieser: 1273 2146
CAS No.: 100-10-7
Molecular Formula: C9H11NO
Formula Weight: 149.19
Flash Point: 164°(327°F)
Physical Form: Crystalline powder
Sensitivity: Air and Light Sensitive
Appearance: White to pale yellow
Odor: Strong
Melting Point: 73-75°
Boiling Point: 176-177°/17mm
Solubility: Slightly soluble in water. Soluble in alcohol, ether, chloroform, acetic acid
Beilstein Reference: 606802
Merck Reference: 14,3230
MDL No.: MFCD00003381
Formula:
(CH₃)₂NC₆H₄CHO MW: 149.19 g/mol Boiling Pt: 176-177 Melting Pt: 72 Density: 1.0254 g/cm³ (99.9 °C) Flash Pt: 164 °C (327 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003381 CAS Number: 100-10-7 EINECS: 202-819-0 Merck Index: 14,3230 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...