Methyl red ACS

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

36682-14 36682-22
CAAA36682-14EA 64.43 CAD
CAAA36682-14 CAAA36682-22
Methyl red ACS
Methyl red

pH indicator in pH range 4.2 - 6.3


CAS No.: 493-52-7 Molecular Formula: C15H15N3O2 Formula Weight: 269.30 Physical Form: Crystalline Appearance: Dark red Melting Point: 179-182° Beilstein Reference: 750102 Merck Reference: 14,6119 MDL No.: MFCD00002425


pH indicator in pH range 4.2 - 6.3

Formula: C₁₅H₁₅N₃O₂
Melting Pt: 179…182 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
EINECS: 207-776-1
Merck Index: 14,6119

Order Now

Specification Test Results

Melting point (range) 179-182°
Clarity of alcohol solution Pass test
Visual transition interval from pH 4.2 (pink) to 6.2 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR