Titanium ≥99.99% (metals basis), powder -325 mesh, dehydrided

Supplier: Thermo Scientific Chemicals
Danger

36679-14 36679-22
CAAA36679-14EA 297.04 CAD
CAAA36679-14 CAAA36679-22
Titanium ≥99.99% (metals basis), powder -325 mesh, dehydrided
Titanium

MDL: MFCD00011264


Purity: 99.99% CAS No.: 7440-32-6 Odor: Odorless UN No.: 2546 MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2 II
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR