Titanium ≥99.99% (metals basis), powder -325 mesh, hydrided

Supplier: Thermo Scientific Chemicals
Danger

36678-14 36678-22
CAAA36678-14EA 221.2 CAD
CAAA36678-14 CAAA36678-22
Titanium ≥99.99% (metals basis), powder -325 mesh, hydrided
Titanium

MDL: MFCD00011264


Purity: 99.99% CAS No.: 7440-32-6 Odor: Odorless UN No.: 2546 MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-726-8
UN: 2546
ADR: 4.2 II
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR