4-(Methylamino)phenol sulfate 99.0-101.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36674-18
36674-30
CAAA36674-18EA
57.51
CAD
CAAA36674-18
CAAA36674-30
4-(Methylamino)phenol sulfate 99.0-101.5% ACS
4-(Methylamino)phenol sulfate
MDL: MFCD00013140
Beilstein Registry No.: 3919382
Purity: 99.0-101.5%
CAS No.: 55-55-0
Molecular Formula: C14H18N2O2.H2SO4
Formula Weight: 344.38
Physical Form: Powder
Sensitivity: Air and Light Sensitive
Appearance: White to pale cream
Melting Point: ca. 260° dec.
Beilstein Reference: 3919382
Merck Reference: 14,6017
UN No.: 2811
MDL No.: MFCD00013140
Formula:
C₁₄H₂₀N₂O₆S Boiling Pt: 227 °C (1013 hPa) Melting Pt: ∼. 260 °C (dec.) Density: 1.434 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013140 CAS Number: 55-55-0 EINECS: 200-237-1 UN: 2811 ADR: 6.1,III Merck Index: 14,6017 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...