Methyl-p-benzoquinone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylcyclohexa-2,5-diene-1,4-dione, Methylquinone, Methyl-1,4-benzoquinone

36670-09 36670-18
CAAA36670-09EA 65.73 CAD
CAAA36670-09 CAAA36670-18
Methyl-p-benzoquinone 98%
Methyl-p-benzoquinone

MDL: MFCD00001603


Purity: 98% CAS No.: 553-97-9 Molecular Formula: C7H6O2 Formula Weight: 122.12 Physical Form: Crystalline Appearance: Yellowish green Melting Point: 67-70° MDL No.: MFCD00001603

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 187
Melting Pt: 67…70 °C
Density: 1.08 g/cm³ (75.5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001603
CAS Number: 553-97-9
EINECS: 209-056-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR