Silicon dioxide 15% in water, colloidal dispersion

Supplier: Thermo Scientific Chemicals
Warning

36669-A1 36669-A7
CAAA36669-A1EA 96.54 CAD
CAAA36669-A1 CAAA36669-A7
Silicon dioxide 15% in water, colloidal dispersion
Silicium dioxide

High-temperature binders, anti-slip coatings for paper, petrochemical catalyst support and binder, polishing agent, abrasion resistant coatings


CAS No.: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.09 Physical Form: 0.004 Micron Particles, in Liquid, S.A. 650m2/g Appearance: Colorless


High-temperature binders, anti-slip coatings for paper, petrochemical catalyst support and binder, polishing agent, abrasion resistant coatings

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR