Methyl red hydrochloride ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(4-Dimethylaminophenylazo)benzoic acid hydrochloride
36668-14
36668-22
CAAA36668-14EA
55.46
CAD
CAAA36668-14
CAAA36668-22
Methyl red hydrochloride ACS
Methyl red hydrochloride
MDL: MFCD00012614
CAS No.: 63451-28-5
Molecular Formula: C15H15N3O2.HCl
Formula Weight: 305.77
Physical Form: Crystalline
Melting Point: 175° dec.
MDL No.: MFCD00012614
Formula:
C₁₅H₁₆ClN₃O₂ MW: 305.76 g/mol Melting Pt: 175 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012614 CAS Number: 63451-28-5 EINECS: 264-190-9 |
Specification Test Results
Clarity of alcohol solution | Pass test |
Visual transition interval | pH 4.2 (pink) to 6.2 (yellow) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...