Mercury(II) telluride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
36666-04
36666-09
CAAA36666-04EA
211.72
CAD
CAAA36666-04
CAAA36666-09
Mercury(II) telluride ≥99.999% (metals basis)
Mercury (II) telluride
MDL: MFCD00016133
Purity: 99.999%
CAS No.: 12068-90-5
Molecular Formula: HgTe
Formula Weight: 328.19
Physical Form: 3-12mm Fused Lump
Density: 8.63
Appearance: Gray to black
Odor: Odorless
Melting Point: 673°
UN No.: 2025
MDL No.: MFCD00016133
Formula:
HgTe Melting Pt: 673 °C Density: 8.63 Storage Temperature: Ambient |
MDL Number:
MFCD00016133 CAS Number: 12068-90-5 EINECS: 235-108-9 UN: 2025 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...