Citric acid, anhydrous ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
36664-22
36664-36
36664-A3
CAAA36664-22EA
63.25
CAD
CAAA36664-22
CAAA36664-36
CAAA36664-A3
Citric acid, anhydrous ≥99.5% ACS
Citric acid
Vitamin C
Purity: 99.5+%
CAS No.: 77-92-9
Molecular Formula: C6H8O7
Formula Weight: 192.13
Physical Form: Crystalline
Density: 1.665
pH: 1,6
Melting Point: 153-154°
Beilstein Reference: 782061
Merck Reference: 14,2326
MDL No.: MFCD00011669
Vitamin C
Formula:
HOC(COOH)(CH₂COOH)₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 153…154 °C Density: 1.665 Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 201-069-1 REACH: 01-2119457026-42-0000 Merck Index: 14,2326 |
|
Specification Test Results
Insoluble matter | 0.005% |
Residue after ignition | 0.02% |
Cl | 0.001% |
C2O3 | Pass test (limit about 0.05%) |
PO3 | 0.001% |
SO3 | 0.002% |
Fe | 3 ppm |
PB | 2 ppm |
Substances carbonizable by hot sulfuric acid | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...