Glycerine ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36646-AP
36646-K2
36646-K7
36646-M1
CAAA36646-APEA
108.23
CAD
CAAA36646-AP
CAAA36646-K2
CAAA36646-K7
CAAA36646-M1
Glycerine ≥99.5% ACS
Glycerine
MDL: MFCD00004722
Beilstein Registry No.: 635685
Purity: 99.5+%
CAS No.: 56-81-5
Molecular Formula: C3H8O3
Formula Weight: 92.09
Flash Point: 160°(320°F)
Physical Form: Viscous liquid
Density: 1.26
Refractive Index: 1.474
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Odorless
Melting Point: 18°
Boiling Point: 182°/20mm
Beilstein Reference: 635685
Merck Reference: 14,4484
MDL No.: MFCD00004722
Formula:
HOCH₂CH(OH)CH₂OH Boiling Pt: 182 °C (20 mmHg) Melting Pt: 18 °C Density: 1.26 Flash Pt: 160 °C (320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 14,4484 |
|
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.005% |
Neutrality | Pass test |
Chlorinated compounds (as Cl) | 0.003% |
Sulfate | 0.001% |
Acrolein and glucose | Pass test |
Fatty acid esters (as butyric acid) | 0.05% |
Substances darkened by sulfuric acid P.T | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...