Isobutanol 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
MDL: MFCD00004740
Beilstein Registry No.: 1730878
Purity: 99+%
CAS No.: 78-83-1
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 0.802
Refractive Index: 1.3960
Appearance: Colorless
Odor: Sweet
pH: 7
Melting Point: -108°
Boiling Point: 107-108°
Beilstein Reference: 1730878
Merck Reference: 14,5131
UN No.: 1212
MDL No.: MFCD00004740
Formula:
(CH₃)₂CHCH₂OH MW: 74.12 g/mol Boiling Pt: 107.9 Melting Pt: -108 Density: 0.802 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004740 CAS Number: 78-83-1 EINECS: 201-148-0 UN: 1212 ADR: 3 III Merck Index: 14,5131 |
|
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.001% |
Solubility in water | Pass test |
Titratable acid | 0.0005meq/g |
H2O | 0.1% |
Carbonyl compounds | 0.01 % (butyraldehyde) and 0.02 % (2-butanone) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...