Propylamine 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
36635-AK
36635-K2
CAAA36635-AKEA
63.52
CAD
CAAA36635-AK
CAAA36635-K2
Propylamine 99+%
Propylamine
MDL: MFCD00008205
Beilstein Registry No.: 1098243
Notes: Miscible with water, alcohol, ether
Purity: 99+%
CAS No.: 107-10-8
Molecular Formula: CH3CH2CH2NH2
Formula Weight: 59.11
Flash Point: -12°(10°F)
Physical Form: Liquid
Density: 0.719
Refractive Index: 1.389
Sensitivity: Air Sensitive
Appearance: Colorless
Odor: Amine-like
pH: 12.6
Melting Point: -83°
Boiling Point: 48-49°
Beilstein Reference: 1098243
Merck Reference: 14,7843
UN No.: 1277
MDL No.: MFCD00008205
Formula:
H₃CCH₂CH₂NH₂ Boiling Pt: 48…49 °C Melting Pt: –83 °C Density: 0.719 Flash Pt: –12 °C (10 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008205 CAS Number: 107-10-8 EINECS: 203-462-3 UN: 1277 ADR: 3,II Merck Index: 14,7843 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...