Cadmium(II) chloride, anhydrous ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cadmium chloride

36629-22 36629-36
CAAA36629-22EA 143.31 CAD
CAAA36629-22 CAAA36629-36
Cadmium(II) chloride, anhydrous ≥99.0% ACS
Cadmium (II) chloride

Preparation of cadmium sulfide, as analytical lab reagent, photography, manufacturing of special mirrors, ice-nucleating agent, lubricant, to absorb H2S in sulfide analysis


Purity: 99.0% min CAS No.: 10108-64-2 Molecular Formula: CdCl2 Formula Weight: 183.32 Physical Form: Powder Density: 4.047 Melting Point: 568° Boiling Point: 960° Solubility: Freely soluble in water. Soluble in acetone. Slightly soluble in methanol. Insoluble in ether Merck Reference: 14,1617 UN No.: 2570 MDL No.: MFCD00010916


Preparation of cadmium sulfide, as analytical lab reagent, photography, manufacturing of special mirrors, ice-nucleating agent, lubricant, to absorb H2S in sulfide analysis

Formula: CdCl₂
MW: 183.32 g/mol
Boiling Pt: 967
Melting Pt: 565
Density: 4.047
Storage Temperature: Ambient
MDL Number: MFCD00010916
CAS Number: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1 III
Merck Index: 14,1617

Order Now

Specification Test Results

Insoluble matter 0.01%
Oxidizing substances (as NO3) 0.003%
SO3 0.01%
NH3 0.01%
Cu 0.001%
Fe 0.001%
PB 0.005%
Ca 0.01%
K 0.02%%
NA 0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR