4,4'-Bi-o-toluidine dihydrochloride ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Toluidine dihydrochloride, o-Tolidine dihydrochloride

36626-36 36626-14 36626-22
CAAAA36626-36EA 782.1 CAD
CAAAA36626-36 CAAA36626-14 CAAA36626-22
4,4'-Bi-o-toluidine dihydrochloride ACS
4,4'-Bi-o-toluidine dihydrochloride

MDL: MFCD00012960


CAS No.: 612-82-8 Molecular Formula: C14H16N2.2HCl Formula Weight: 285.22 Physical Form: Solid Sensitivity: Air and Light Sensitive Appearance: White to cream Odor: Odorless Melting Point: 300° UN No.: 3077 MDL No.: MFCD00012960

Formula: C₁₄H₁₈Cl₂N₂
MW: 285.22 g/mol
Melting Pt: 130
Storage Temperature: Ambient
MDL Number: MFCD00012960
CAS Number: 612-82-8
EINECS: 210-322-5
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Form Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR