Thiourea ≥99%, REacton® ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36609-18
36609-30
36609-A1
CAAA36609-18EA
64.62
CAD
CAAA36609-18
CAAA36609-30
CAAA36609-A1
Thiourea ≥99%, REacton® ACS
Thiourea
Reagent for organic synthesis
Purity: 99% min.
CAS No.: 62-56-6
Molecular Formula: CH4N2S
Formula Weight: 76.12
Physical Form: Crystalline
Density: 1.405
Appearance: White
Odor: Odorless
Melting Point: 175-178°
Beilstein Reference: 605327
Merck Reference: 14,9367
UN No.: 3077
MDL No.: MFCD00008067
Reagent for organic synthesis
Formula:
NH₂CSNH₂ Melting Pt: 175…178 °C Density: 1.405 Storage Temperature: Ambient |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 EINECS: 200-543-5 UN: 3077 ADR: 9,III Merck Index: 14,9367 |
|
Specification Test Results
Solubility in water | Pass test |
Residue after ignition | 0.1% |
Loss on drying | 0.005 |
Melting point | 174-177° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...