Zinc ≥99.8%, granules ACS -20 mesh

Supplier: Thermo Scientific Chemicals

36602-22 36602-36 36602-A1
CAAA36602-22EA 68.73 CAD
CAAA36602-22 CAAA36602-36 CAAA36602-A1
Zinc ≥99.8%, granules ACS -20 mesh
Zinc

MDL: MFCD00011291


Purity: 99.8% min CAS No.: 7440-66-6 Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Specification Test Results

Fe 0.01% Max.
PB 0.01% Max.
Suitability for determination of As Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR