Soda lime ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
36596-36
36596-A4
CAAA36596-36EA
109.34
CAD
CAAA36596-36
CAAA36596-A4
Soda lime ACS
Soda lime
MDL: MFCD00134094
Notes: Variable mix of sodium hydroxide with calcium oxide or hydroxide
CAS No.: 8006-28-8
Physical Form: 4-8 Mesh
Sensitivity: Air Sensitive and Hygroscopic
Appearance: White
Odor: Odorless
Merck Reference: 14,8569
MDL No.: MFCD00134094
Storage Temperature:
Ambient |
MDL Number:
MFCD00134094 CAS Number: 8006-28-8 UN: 1907 ADR: 8,III Merck Index: 14,8569 |
Specification Test Results
Loss on drying at 200°C | 7% |
Maximum CO2 absorption capacity | 0.19 |
minimum Fines | 0.01 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...