Zinc(II) telluride ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36595-09
36595-18
CAAA36595-09EA
211.72
CAD
CAAA36595-09
CAAA36595-18
Zinc(II) telluride ≥99.998% (metals basis)
Zinc (II) telluride
MDL: MFCD00043079
Purity: 99.998%
CAS No.: 1315-11-3
Molecular Formula: ZnTe
Formula Weight: 192.97
Physical Form: 3-12mm Lump
Density: 6.34
Refractive Index: 3.56
Appearance: Red
Odor: Odorless
Melting Point: 1240°
Merck Reference: 14,10162
MDL No.: MFCD00043079
Formula:
TeZn Melting Pt: 1240 °C Density: 6.34 Storage Temperature: Ambient |
MDL Number:
MFCD00043079 CAS Number: 1315-11-3 EINECS: 215-260-2 Merck Index: 14,10162 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...