Tetraethylammonium hydroxide 35% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetrylammonium hydroxide

36560-22 36560-36 36560-A3
CAAA36560-22EA 99.22 CAD
CAAA36560-22 CAAA36560-36 CAAA36560-A3
Tetraethylammonium hydroxide 35% (w/w) in aqueous solution
Tetraethylammonium hydroxide

MDL: MFCD00009024 Fieser: 1,1138


CAS No.: 77-98-5 Molecular Formula: C8H21NO Formula Weight: 147.26 Physical Form: Liquid Density: 1.023 Refractive Index: 1.4040 Merck Reference: 14,9201 UN No.: 3267 MDL No.: MFCD00009024

Formula: (C₂H₅)₄N(OH)
MW: 147.26 g/mol
Density: 1.023
Storage Temperature: Ambient
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
UN: 3267
ADR: 8,II
Merck Index: 14,9201

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR