Dimethyl sulfoxide-[D₆] (99.9% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulfoxide, Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulfoxide-[²H₆], Dimethyl sulphoxide-d6, Dimethyl sulfoxide-d6

36517-46 036517-48 36517-48
CAAA36517-46EA 109.34 CAD
CAAA36517-46 AA36517-48 CAAA36517-48
Dimethyl sulfoxide-[D₆] (99.9% D) (Isotopic) for NMR spectroscopy
Dimethyl sulphoxide-[D6]

MDL: MFCD00002090


Purity: 99.9% CAS No.: 2206-27-1 Molecular Formula: (CD3)2SO Formula Weight: 84.17 Flash Point: 88°(190°F) Physical Form: Liquid Density: 1.18 Refractive Index: 1.4760 Sensitivity: Hygroscopic Appearance: Colorless Odor: Characteristic Melting Point: 20° Boiling Point: 190° MDL No.: MFCD00002090

Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: 190 °C
Melting Pt: 20 °C
Density: 1.18
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR