p-Toluenesulfonic acid monohydrate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00142137
Beilstein Registry No.: 3568023
Fieser: 1,1172 4,508 5,673 6,597 7,374 8,488 9,471 11,535 12,507
Purity: 97%
CAS No.: 6192-52-5
Molecular Formula: CH3C6H4SO3H.H2O
Formula Weight: 190.22 (172.20anhy)
Flash Point: 150°(302°F)
Physical Form: Crystalline
Density: 1.24
Sensitivity: Hygroscopic
Appearance: White to pink
Odor: Mild
Melting Point: 103-106°(anhy)
Boiling Point: 140°/20mm
Beilstein Reference: 3568023
Merck Reference: 14,9533
UN No.: 2585
MDL No.: MFCD00142137
Formula:
C₇H₈O₃S·1H₂O Boiling Pt: 140 °C (20 mmHg) Melting Pt: 103…106 °C (anhydr.) Density: 1.24 Flash Pt: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00142137 CAS Number: 6192-52-5 EINECS: 203-180-0 UN: 2585 ADR: 8,III Merck Index: 14,9533 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...