Formic acid ≥88% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
36504-AP
36504-AY
36504-K3
CAAA36504-APEA
141.99
CAD
CAAA36504-AP
CAAA36504-AY
CAAA36504-K3
Formic acid ≥88% ACS
Formic acid
MDL: MFCD00003297
Beilstein Registry No.: 1209246
Fieser: 1,404 8,232 9,226 11,243 13,137 18,163 19,148 20,168 21,193
CAS No.: 64-18-6
Molecular Formula: CH2O2
Formula Weight: 46.03
Flash Point: 50°(156°F)
Physical Form: Liquid
Density: 1.220
Refractive Index: 1.3710
Sensitivity: Hygroscopic
Appearance: Colorless
Melting Point: 7-9°
Boiling Point: 100-101°
Beilstein Reference: 1209246
Merck Reference: 14,4241
UN No.: 1779
MDL No.: MFCD00003297
Formula:
CH₂O₂ Boiling Pt: 100…101 °C Melting Pt: 7…9 °C Density: 1.220 Flash Pt: 50 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8,II Merck Index: 14,4241 |
|
Specification Test Results
Color (APHA) | 15 |
Dilution test | Pass Test |
Evaporation residue | 0.002 |
CH3COOH | 0.004 |
NH3 | 0.00005 |
Cl | 0.001% |
SO3 | Pass test |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...