Dichloromethane-[D₂] (99.9% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dichloromethane-d2, Methylene chloride-D2, Dichloromethane-[²H₂], Dideuteromethylene chloride, Methylene chloride-d2

36500-03 36500-06
CAAA36500-03EA 50.24 CAD
CAAA36500-03 CAAA36500-06
Dichloromethane-[D₂] (99.9% D) (Isotopic) for NMR spectroscopy
Dichloromethane-[D2]

MDL: MFCD00000882


Purity: 99.9% CAS No.: 1665-00-5 Molecular Formula: CD2Cl2 Formula Weight: 86.95 Physical Form: Liquid Density: 1.35 UN No.: 1593 MDL No.: MFCD00000882

Formula: CD₂Cl₂
MW: 86.92 g/mol
Boiling Pt: 40
Melting Pt: –97 °C
Density: 1.35
Storage Temperature: Ambient
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR