Gum arabic (total ash max 4%)

Supplier: Thermo Scientific Chemicals

Synonyms: Acacia gumAcacia

36499-36 36499-A3 36499-36 36499-A3
CAAA36499-36EA 111 CAD
AA36499-A3 AA36499-36 CAAA36499-36 CAAA36499-A3
Gum arabic (total ash max 4%)
Gum arabic

Gum arabic (Acacia) is a heterogeneous material having both hydrophilic and hydrophobic affinities.


It is used in textiles, ceramics, lithography, cosmetics and pharmaceuticals. In the food industry, acacia is mainly used as confectionery, bakery, dairy, drink, and as a microencapsulating agent.

Density: 1.4
Flash Pt: >250 °C
MDL Number: MFCD00081264
CAS Number: 9000-01-5
Merck Index: 14,14

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR