Sodium tungstate dihydrate 99.0-101.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36489-22
36489-36
CAAA36489-22EA
138.09
CAD
CAAA36489-22
CAAA36489-36
Sodium tungstate dihydrate 99.0-101.0% ACS
Sodium wolframate dihydrate
MDL: MFCD00149190
Fieser: 13,145 15,295
Purity: 99.0-101.0%
CAS No.: 10213-10-2
Molecular Formula: Na2WO4.2H2O
Formula Weight: 329.86 (293.83anhy)
Physical Form: Powder
Density: 3.250
Appearance: White
Odor: Odorless
Melting Point: 100° -2H2O
Merck Reference: 14,8698
MDL No.: MFCD00149190
Formula:
Na₂WO₄·2H₂O Melting Pt: 100 °C (-2H₂O) Density: 3.250 Storage Temperature: Ambient |
MDL Number:
MFCD00149190 CAS Number: 10213-10-2 EINECS: 236-743-4 Merck Index: 14,8698 |
Specification Test Results
Insoluble matter | 0.01% |
Titratable free base | 0.02meq/gmeq/g |
Cl | 0.005% |
Mo | 0.001% |
SO3 | 0.01% |
Heavy metals and iron (as Pb) | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...