Lecithin, refined

Supplier: Thermo Scientific Chemicals

36486-30 36486-A1
CAAA36486-30EA 143.23 CAD
CAAA36486-30 CAAA36486-A1
Lecithin, refined
Lecithin

Lecithin is used as an emulsifying and stabilizing agent in the food, pharmaceutical, and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacturing of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.


CAS No.: 8002-43-5 Physical Form: Powder Odor: Odorless Beilstein Reference: 5209585 Merck Reference: 14,5428 MDL No.: MFCD00147406


Caution: RUO – Research Use Only

Density: 1.0305 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00147406
CAS Number: 8002-43-5
EINECS: 232-307-2
Merck Index: 14,5428

Order Now

Specification Test Results

Appearance (Color) Light tan to medium yellow
Form Powder
Assay (unspecified) Hexane insoluble 0.3%, max
Acid Value <35 mg KOH/g
Residual water < 1%
Peroxide content 5 meq /kg, max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR