Sodium carbonate monohydrate ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36485-22
36485-36
36485-A3
CAAA36485-22EA
60.51
CAD
CAAA36485-22
CAAA36485-36
CAAA36485-A3
Sodium carbonate monohydrate ≥99.5% ACS
Sodium carbonate monohydrate
MDL: MFCD00149177
Purity: 99.5% min
CAS No.: 5968-11-6
Molecular Formula: Na2CO3.H2O
Formula Weight: 124.01 (105.99anhy)
Physical Form: Granular
Density: 2.250
Appearance: Colorless to white
Odor: Odorless
Melting Point: 100° -H2O
Merck Reference: 14,8596
MDL No.: MFCD00149177
Formula:
Na₂CO₃·1H₂O Melting Pt: 100 °C (-H₂O) Density: 2.250 Storage Temperature: Ambient |
MDL Number:
MFCD00149177 CAS Number: 5968-11-6 EINECS: 207-838-8 Merck Index: 14,8596 |
Specification Test Results
Insoluble matter | 0.01% |
Loss on drying at 150° | 13.0-15.0% |
Cl | 0.001% |
PO3 | 5 ppm |
SiO2 | 0.005% |
Sulfur compounds (asSO3) | 0.004% |
Ca | 0.0003 |
MG | 0.00005 |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5ppm ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...