Copper ≥99.9% (metals basis), shot ACS ≤ 6 mm (0.2 in)

Supplier: Thermo Scientific Chemicals

36482-22 36482-36
CAAA36482-22EA 88.95 CAD
CAAA36482-22 CAAA36482-36
Copper ≥99.9% (metals basis), shot ACS ≤ 6 mm (0.2 in)
Copper

MDL: MFCD00010965


Purity: 99.9+% CAS No.: 7440-50-8 Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Specification Test Results

Matter insoluble in dilute nitric acid 0.02% Max.
Antimony and tin (as Sn) 0.01 Max.
As 5 ppm Max.
Fe 0.005% Max.
PB 0.005% Max.
Mn 0.001% Max.
P 0.001% Max.
Ag 0.002% Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR