Dimethyl sulfoxide ≥99.9% ACS
Supplier: Thermo Scientific Chemicals
Warning
|
36480-AP
36480-K2
36480-K7
36480-M1
36480-M6
CAAA36480-APEA
133.54
CAD
CAAA36480-AP
CAAA36480-K2
CAAA36480-K7
CAAA36480-M1
CAAA36480-M6
Dimethyl sulfoxide ≥99.9% ACS
MDL: MFCD00002089
Beilstein Registry No.: 506008
Fieser: 1,296 14,150 15,146 16,149 17,127 18,149 20,154 21,180
Purity: 99.9% min
CAS No.: 67-68-5
Molecular Formula: C2H6OS
Formula Weight: 78.13
Flash Point: 87°(189°F)
Physical Form: Clear liquid
Density: 1.101
Refractive Index: 1.4790
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Mild
Melting Point: 18.4°
Boiling Point: 189°
Beilstein Reference: 506008
Merck Reference: 14,3259
MDL No.: MFCD00002089
Formula:
(CH₃)₂SO Boiling Pt: 189 °C Melting Pt: 18.4 °C Density: 1.101 Flash Pt: 87 °C (189 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 14,3259 |
|
Specification Test Results
Evaporation residue | 0.01% |
Titratable acid | 0.001meq/gmeq/g |
H2O | 0.1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...