Niobium(V) oxide ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diniobium pentaoxide, Niobium pentoxide
36478-14
36478-22
CAAA36478-14EA
133.67
CAD
CAAA36478-14
CAAA36478-22
Niobium(V) oxide ≥99.95% (metals basis)
Niobium (V) oxide
For vacuum deposition
Purity: 99.95%
CAS No.: 1313-96-8
Molecular Formula: Nb2O5
Formula Weight: 265.81
Physical Form: 3-12mm Sintered Lump
Density: 4.6
Melting Point: 1520°
Merck Reference: 14,6559
MDL No.: MFCD00011128
For vacuum deposition
Formula:
Nb₂O₅ Melting Pt: 1520 °C Density: 4.6 Storage Temperature: Ambient |
MDL Number:
MFCD00011128 CAS Number: 1313-96-8 EINECS: 215-213-6 Merck Index: 14,6559 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...