o-Xylene-[D₁₀] (98% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Xylene-[²H₁₀]1,2-Dimethylbenzene-D10

36472-06 36472-06
CAAA36472-06EA 401.32 CAD
CAAA36472-06 AA36472-06
o-Xylene-[D₁₀] (98% D) (Isotopic) for NMR spectroscopy
o-Xylene-[D10]
Formula: C₆D₄(CD₃)₂
MW: 116.09 g/mol
Boiling Pt: 143…145 °C
Melting Pt: –25 °C
Density: 0.953
Flash Pt: 30 °C
MDL Number: MFCD00037698
CAS Number: 56004-61-6
UN: 1307
ADR: 3,III
Merck Index: 14,10081

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR