Pyridine-[D₅] (99.5% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
36471-04
36471-09
CAAA36471-04EA
75.84
CAD
CAAA36471-04
CAAA36471-09
Pyridine-[D₅] (99.5% D) (Isotopic) for NMR spectroscopy
Pyridine-[D5]
MDL: MFCD00044639
Beilstein Registry No.: 114377
Notes: NMR Solvent
Purity: 99.5%
CAS No.: 7291-22-7
Molecular Formula: C5D5N
Formula Weight: 84.14
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 1.05
Refractive Index: 1.506
Odor: Unpleasant
Melting Point: -42°
Boiling Point: 115°
Beilstein Reference: 114377
UN No.: 1282
MDL No.: MFCD00044639
Formula:
C₅D₅N Boiling Pt: 115 °C Melting Pt: –42 °C Density: 1.05 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044639 CAS Number: 7291-22-7 EINECS: 203-720-2 UN: 1282 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...