Chloroform-[D₁] (99.8% D) (Isotopic) +1% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
36470-14
36470-36
36470-22
CAAAA36470-14EA
63.04
CAD
CAAAA36470-14
CAAAA36470-36
CAAA36470-22
Chloroform-[D₁] (99.8% D) (Isotopic) +1% v/v TMS for NMR spectroscopy
Chloroform-[D1]
MDL: MFCD00000827
Purity: 99.8%
CAS No.: 865-49-6
Molecular Formula: CDCl3
Formula Weight: 120.38
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.50
Refractive Index: 1.4445
Sensitivity: Moisture and Light Sensitive
Appearance: Colorless
Melting Point: -64°
Boiling Point: 62°
UN No.: 1888
MDL No.: MFCD00000827
Formula:
CDCl₃ Boiling Pt: 62 °C Melting Pt: –64 °C Density: 1.50 Storage Temperature: Refrigerator |
MDL Number:
MFCD00000827 CAS Number: 865-49-6 EINECS: 212-742-4 UN: 1888 ADR: 6.1,III |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...