Acetic acid-[D₄] (99.5% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethanoic acid-D4, Acetic-D3 acid-D, Acetic-d3 acid-d, Acetic acid-[²H₄], Tetradeuteroacetic acid, Acetic acid-d₄
36469-06
36469-09
CAAA36469-06EA
92.75
CAD
CAAA36469-06
CAAA36469-09
Acetic acid-[D₄] (99.5% D) (Isotopic) for NMR spectroscopy
Acetic acid-[D4]
MDL: MFCD00051051
Notes: NMR Solvent
Purity: 99.5%
CAS No.: 1186-52-3
Molecular Formula: CD3CO2D
Formula Weight: 64.08
Physical Form: Liquid
Density: 1.12
Melting Point: 17°
Boiling Point: 118°
UN No.: 2789
MDL No.: MFCD00051051
Formula:
D₃CCOOD Boiling Pt: 118 °C Melting Pt: 17 °C Density: 1.12 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051051 CAS Number: 1186-52-3 EINECS: 214-693-4 UN: 2789 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...