Hexamethylenetetramine 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

36462-22 36462-36 36462-A3
CAAA36462-22EA 50.78 CAD
CAAA36462-22 CAAA36462-36 CAAA36462-A3
Hexamethylenetetramine 99+% ACS
Methenamine

MDL: MFCD00006895 Beilstein Registry No.: 2018 Fieser: 1,427 2,208 4,243 9,234 18,178 19,164 20,185


Purity: 99+% CAS No.: 100-97-0 Molecular Formula: C6H12N4 Formula Weight: 140.19 Flash Point: 250°(482°F) Physical Form: Solid Density: 1.33 Sensitivity: Hygroscopic Appearance: White Melting Point: 280° subl. Beilstein Reference: 2018 Merck Reference: 14,5966 UN No.: 1328 MDL No.: MFCD00006895

Formula: C₆H₁₂N₄
Melting Pt: 280 °C (subl.)
Density: 1.33
Flash Pt: 250 °C (482 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
EINECS: 202-905-8
UN: 1328
ADR: 4.1,III
Merck Index: 14,5966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3ecbb5df5394322c2150525ce6e9f48b5aadde7&vid=04b623f7f333c7cf159016619cf91fc792af9a3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Loss on drying 2%
Residue after ignition 0.1%
Heavy Metals (as Pb) 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR