Sodium bromide ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Warning
|
36444-36
36444-A3
CAAA36444-36EA
141.57
CAD
CAAA36444-36
CAAA36444-A3
Sodium bromide ≥99.0% ACS
Sodium bromide
MDL: MFCD00003475
Fieser: 12,445
Purity: 99.0% min
CAS No.: 7647-15-6
Molecular Formula: NaBr
Formula Weight: 102.89
Physical Form: Crystalline
Density: 3.203
Refractive Index: 1.6412
Sensitivity: Hygroscopic
Melting Point: 755°
Boiling Point: 1390°
Merck Reference: 14,8594
MDL No.: MFCD00003475
Formula:
NaBr Boiling Pt: 1390 °C Melting Pt: 755 °C Density: 3.203 Storage Temperature: Ambient |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 EINECS: 231-599-9 Merck Index: 14,8594 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 5.0-8.8 at 25° |
BrO4 | 0.001% |
Cl | 0.2% |
SO3 | 0.002% |
Ba | 0.002% |
Ca | 0.002% |
MG | 0.001% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...