N,N-Dimethyl-1-naphthylamine ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
36438-AA
36438-AC
CAAA36438-AAEA
57.2
CAD
CAAA36438-AA
CAAA36438-AC
N,N-Dimethyl-1-naphthylamine ≥99%
N,N-Dimethyl-1-naphthylamine
MDL: MFCD00003919
Purity: 99%
CAS No.: 86-56-6
Molecular Formula: C10H7N(CH3)2
Formula Weight: 171.24
Physical Form: Liquid
Density: 1.039
Refractive Index: 1.6230
Appearance: Pale yellow
Boiling Point: 145-147°/12mm
Merck Reference: 14,3252
UN No.: 3082
MDL No.: MFCD00003919
Formula:
C₁₂H₁₃N Boiling Pt: 145…147 °C (12 mmHg) Density: 1.039 Storage Temperature: Ambient |
MDL Number:
MFCD00003919 CAS Number: 86-56-6 EINECS: 201-682-4 UN: 3082 ADR: 9,III Merck Index: 14,3252 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...