Glycine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

36435-30 36435-A1
CAAA36435-30EA 85.08 CAD
CAAA36435-30 CAAA36435-A1
Glycine 99+%
Glycine

Purity: 99+% CAS No.: 56-40-6 Molecular Formula: H2NCH2CO2H Formula Weight: 75.07 Physical Form: Powder Density: 1.595 pH: 5,97 Melting Point: ca 245° dec. Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131

Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: ∼ 245 °C (dec.)
Density: 1.595
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 14,4491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR