Gallic acid monohydrate 98+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36417-22
36417-36
CAAA36417-22EA
106.02
CAD
CAAA36417-22
CAAA36417-36
Gallic acid monohydrate 98+% ACS
Gallic acid monohydrate
MDL: MFCD00149098
Purity: 98+%
CAS No.: 5995-86-8
Molecular Formula: C7H6O5.H2O
Formula Weight: 188.14 (170.12anhy)
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to cream
Melting Point: 250° dec.
Merck Reference: 14,4345
MDL No.: MFCD00149098
Formula:
C₇H₆O₅·1H₂O Melting Pt: 250 °C (dec.) Flash Pt: 250 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00149098 CAS Number: 5995-86-8 EINECS: 205-749-9 Merck Index: 14,4345 |
Specification Test Results
Insoluble matter | 0.01% |
Residue after ignition | 0.05% |
SO3 | 0.02% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...