Hydroxylammonium chloride ≥96% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hydroxylamine HCl, Hydroxylamine hydrochloride
A monoamine oxidase inhibitor; inhibits platelet aggregation
Purity: 96+%
CAS No.: 5470-11-1
Molecular Formula: H3NO.HCl
Formula Weight: 69.49
Physical Form: Solid
Density: 1.67
Appearance: White
Odor: Acidic
pH: 3.6
Melting Point: 152° dec.
Solubility: Soluble in water, alcohol, methanol, glycerol
Beilstein Reference: 3539763
Merck Reference: 14,4828
UN No.: 2923
MDL No.: MFCD00051089
A monoamine oxidase inhibitor; inhibits platelet aggregation
Formula:
NH₂OH·HCl MW: 69.49 g/mol Boiling Pt: 306 °C (1013 hPa) Melting Pt: 152 °C (dec.) Density: 1.67 Storage Temperature: Ambient |
MDL Number:
MFCD00051089 CAS Number: 5470-11-1 EINECS: 226-798-2 UN: 2923 ADR: 8,III Merck Index: 14,4828 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...