L(+)-Tartaric acid ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
Useful as a resolving agent
CAS No.: 87-69-4
Molecular Formula: C4H6O6
Formula Weight: 150.09
Physical Form: Granular
Density: 1.760
Odor: Odorless
pH: 2.1
Melting Point: 168-170°
Solubility: Soluble in water, methanol, ethanol, glycerol
Beilstein Reference: 1725147
Merck Reference: 14,9070
MDL No.: MFCD00064207
Useful as a resolving agent
Formula:
C₄H₆O₆ Boiling Pt: 399 °C (1013 hPa) Melting Pt: 168…170 °C Density: 1.760 Flash Pt: 150 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00064207 CAS Number: 87-69-4 EINECS: 201-766-0 REACH: 01-2119537204-47 Merck Index: 14,9070 |
|
Specification Test Results
Insoluble matter | 0.005 |
Residue after ignition | 0.02 |
Cl | 0.001 |
C2O3PO3 | Pass Test (limit about 0.1%) |
PO3 | 0.001% |
Sulfur compounds (as S) | 0.002% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...